Skip to Content
Merck
All Photos(2)

Key Documents

146755

Millipore

Tryptic Soy Agar - Ready-to-use Settle Plates

IsoBag®, Lecithin, Sodium thiosulfate, Histidine, Tween®, 190 mm alpha-port, ICR plus lockable Plates, sterile; irradiated, plate diam. 90 mm, suitable for air monitoring

Synonym(s):

IsoBag® Tryptic Soy Agar Settle Plus, TSA, Soybean Casein Digest Agar

Sign Into View Organizational & Contract Pricing

Select a Size

80 EA
€1,570.00

€1,570.00


Estimated to ship on31 March 2025Details


Request more information

Select a Size

Change View
80 EA
€1,570.00

About This Item

UNSPSC Code:
41106217
NACRES:
NA.74

€1,570.00


Estimated to ship on31 March 2025Details


Request more information

material

polystyrene plate

Quality Level

Agency

EP
USP

sterility

sterile; irradiated

form

plate (ready-to-use)

feature

rapid transfer port

composition

Histidine
Lecithin
Sodium thiosulfate
Tween®

packaging

pkg of 80 plates

manufacturer/tradename

IsoBag®

plate diam.

90 mm

pH

7.3 ( in H2O)

application(s)

air monitoring
pharmaceutical

compatibility

for use with MAS-100®

shipped in

ambient

storage temp.

15-25°C

suitability

bacteria
fungi

General description

The IsoBag® TSA Settle Plus is designed for quick and convenient transfer of culture media used for air monitoring in isolators or RAB systems. The Beta bag consists of a sterile polyurethane DPTE Bag compatible with 190 mm Alpha Ports. The bag contains 80 lockable 90 mm settle plates filled with Tryptic Soy Agar and supplemented with pyruvate to provide an efficient neutralization of hydrogen peroxide, Lecithin, polysorbate (Tween®) 80, histidine and sodium thiosulfate to inactivate disinfectant residues. In addition, the plate model (plus or “+”) is supplied with a lockable lid for safe transport after sampling and for aerobic and anaerobic incubation of the plates .

Application

The IsoBag® TSA Settle plus is designed for quick and convenient transfer of culture media needed for air monitoring in isolators or RABS systems

Legal Information

ISOBAG is a registered trademark of Merck KGaA, Darmstadt, Germany
MAS-100 is a registered trademark of MBV AG
TWEEN is a registered trademark of Croda International PLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

IsoBag® ease the environmental monitoring workflow in aseptic production isolator

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service