Skip to Content
Merck
All Photos(3)

Key Documents

1.46814

Millipore

Tryptic Soy Agar - Ready-to-use Settle Plates

IsoBag®, 190 mm alpha-port, ICR plus lockable Plates, sterile; irradiated, plate diam. 90 mm, suitable for air monitoring

Synonym(s):

IsoBag® Tryptic Soy Agar Settle, TSA, Soybean Casein Digest Agar

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€1,560.00

€1,560.00


In StockDetails


Request more information

Select a Size

Change View
1 EACH
€1,560.00

About This Item

UNSPSC Code:
41106217
NACRES:
NA.74

€1,560.00


In StockDetails


Request more information

material

polystyrene plate

Agency

according to EP
according to USP

sterility

sterile; irradiated

form

plate

shelf life

9 mo.

feature

rapid transfer port

composition

Soybean Casein Digest Agar

manufacturer/tradename

IsoBag®

packaging

triple packed of

technique(s)

Rapid Sterility Testing: suitable

plate diam.

90 mm

pH

7.1-7.5

solubility

water: soluble

application(s)

air monitoring
environmental
pharmaceutical

compatibility

for use with MAS-100®

storage temp.

15-25°C

suitability

bacteria
molds
yeasts

Looking for similar products? Visit Product Comparison Guide

General description

The IsoBag® TSA Settle is designed for quick and convenient transfer of culture media for air monitoring into isolators or RABS systems. The IsoBag® TSA Settle is a sterile polyurethane DPTE Beta Bag, compatible with 190 mm Getinge DPTE Alpha Ports, containing 8 stacks of 10 lockable 90 mm settle plates (article 146001) each and 4x5 single bagged, pre-sterilized zip bags for safe transport of used plates. The filled IsoBag® product is double bagged and irradiated at a dose of 10-20 kGy.The formulation of the Tryptic Soy Agar contains a combination of casein and soy peptones that renders the medium nutritious by supplying organic nitrogen, particularly amino–acids, and longer-chained peptides. The medium is supplemented with pyruvate to provide an efficient neutralization of hydrogen peroxide.

Application

The IsoBag® TSA Settle is a sterile polyurethane DPTE Beta Bag designed for quick and convenient transfer of culture media for air monitoring into isolators or RABS systems

Legal Information

ISOBAG is a registered trademark of Merck KGaA, Darmstadt, Germany
MAS-100 is a registered trademark of MBV AG

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

IsoBag® ease the environmental monitoring workflow in aseptic production isolator

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service