Skip to Content
Merck
All Photos(1)

Key Documents

14-536

Sigma-Aldrich

MAP Kinase 2/Erk2 Protein, inactive, Human, 50 g

Unactive, N-terminal GST-tagged, recombinant human full length MAP Kinase 2, for use in Kinase Assays.

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€578.00

€578.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
50 μG
€578.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

€578.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in E. coli

mol wt

Mw 67.8 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

shipped in

dry ice

General description

N-terminal GST-tagged, recombinant human full length MAP Kinase 2
Product Source: Expressed in E. coli

Application

Research Category
Metabolism

Inflammation & Immunology
Research Sub Category
Obesity

Metabolic Disorders

Osteoporosis

Arthritis

Biochem/physiol Actions

Protein Target: MAPK2
Target Sub-Family: CMGC

Quality

routinely evaluated by phosphorylation of MBP substrate

Physical form

Glutathione agarose affinity chromatography

Storage and Stability

6 months at -20°C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoyun Wang et al.
Scientific reports, 6, 28260-28260 (2016-06-16)
Although the translational function of tRNA has long been established, extra translational functions of tRNA are still being discovered. We previously developed a computational method to systematically predict new tRNA-protein complexes and experimentally validated six candidate proteins, including the mitogen-activated
Ivana Yen et al.
Cancer cell, 34(4), 611-625 (2018-10-10)
Targeting KRAS mutant tumors through inhibition of individual downstream pathways has had limited clinical success. Here we report that RAF inhibitors exhibit little efficacy in KRAS mutant tumors. In combination drug screens, MEK and PI3K inhibitors synergized with pan-RAF inhibitors

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service