Skip to Content
Merck
All Photos(1)

Key Documents

14-550-M

Sigma-Aldrich

MAP Kinase 2/Erk2 Protein, active, Human, 10 µg

Active, N-terminal GST tagged, recombinant, full-length, human MAP Kinase 2/Erk2. Activated with MEK1 (Cat. # 14-429). For use in Kinase Assays.

Synonym(s):

Erk2 kinase

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€578.00
250 μG
€3,770.00

€578.00


Estimated to ship on24 April 2025Details


Request a Bulk Order

Select a Size

Change View
10 μG
€578.00
250 μG
€3,770.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

€578.00


Estimated to ship on24 April 2025Details


Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in E. coli

mol wt

Mw 67.8 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

General description

N-terminal GST tagged, recombinant, full-length, human MAP Kinase 2/Erk2, Activated with MEK1 (14-429).

Packaging

Also available in 125μg size, please inquire for pricing and availability.

Quality

Used to phosphorylate MBP (13-104)

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanping Li et al.
The Journal of biological chemistry, 288(38), 27646-27657 (2013-07-31)
The small G protein Ras regulates proliferation through activation of the mitogen-activated protein (MAP) kinase (ERK) cascade. The first step of Ras-dependent activation of ERK signaling is Ras binding to members of the Raf family of MAP kinase kinase kinases

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service