Skip to Content
Merck
All Photos(1)

Documents

1.44194

Millipore

RCS® High Flow Touch Microbial Air Sampler

Impaction air sampler based on centrifugal force

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103403
NACRES:
NA.74

material

polycarbonate hardware

product line

RCS®

feature

model (portable)
portable

packaging

pkg of 1 kit

manufacturer/tradename

RCS®

weight

1.5 kg

application(s)

air monitoring
cosmetics
food and beverages
pharmaceutical

compatibility

for use with Agar Strips

storage temp.

room temp

General description

RCS® High Flow Touch is a microbial air sampling system consisting of an instrument and agar strips, for monitoring of ambient air and compressed gases. The RCS® High Flow Touch Microbial Air Sampler employs the principle of centrifugal impaction, whereby air is taken into a rotor and accelerated. The micro-organisms in the air stream are then gently applied to ready-to-use HYCON® Agar Strips which, after a defined incubation period, is evaluated. The instrument is equipped with a high-resolution color touchscreen for convenient and fast handling and the battery capacity to perform more than 35 x 1000 L measurements with one full charging cycle. The instrument is delivered in a transportation case including an USB Stick with RCS® Management software, User manual, Rotor, Protection Cap, Power Supply, Serial RS232 Cable, USB-Adapter, Calibration Certificate and a Quick Start Guide.

Application

The RCS® High Flow Touch delivers reliable reproducible results for applications assessing the quality of ambient air in cleanrooms and isolators for use in pharmaceutical, medical, food and cosmetics industries.

Features and Benefits

Reliable
  • Proven technology using standardized agar media
  • Innovative battery concept with advanced control options
  • Robust instrument that requires minimal service and maintenance
Fast
  • Short sampling times with a flow rate of 100 L/min
  • Convenient programming with an easy touch
Flexible
  • Portable, battery driven and light weight
  • Horizontal and vertical installation, measurement at heights of up to 3 meters
  • User-defined sampling options (e.g. individual volumes, time delay, interval sampling)
User-Friendly
  • High resolution color touchscreen with commonly used functions and symbols
  • Intuitive user interface for user-friendly navigation
  • Innovative software solutions easily integrated
Technology
  • Low impaction speed, turbulence and controlled air stream
  • Even distribution of microorganisms
  • No local drying of the agar
  • Easy disinfection, autoclavable sampling head

Legal Information

HYCON is a registered trademark of Merck KGaA, Darmstadt, Germany
RCS is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Food microbiology testing is a key part of the food safety and quality management. Learn more about the wide range of tests covering pathogens and spoilage organisms.

Ensure microbial air sampling generates precise results complying with international standards. Our systems are easy-to-use and reliable.

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service