Skip to Content
Merck
All Photos(1)

Key Documents

1.44107

Millipore

Hycon® Blank Strip Kit

Empty Agar Strips, for use with RCS® High Flow Touch, suitable for air monitoring

Synonym(s):

Hycon Ready-to-use Agar Strips

Sign Into View Organizational & Contract Pricing

Select a Size

50 UNITS
€134.00

€134.00


Estimated to ship on08 April 2025Details


Request more information

Select a Size

Change View
50 UNITS
€134.00

About This Item

UNSPSC Code:
41113035
NACRES:
NA.74

€134.00


Estimated to ship on08 April 2025Details


Request more information

Quality Level

form

plate

feature

individually wrapped

packaging

pkg of 50 units

manufacturer/tradename

HYCON®

application(s)

air monitoring
cosmetics
food and beverages
pharmaceutical

compatibility

for use with RCS® High Flow Touch

storage temp.

15-25°C

Looking for similar products? Visit Product Comparison Guide

General description

The Blank Strip Kit consists of empty, sealed strips manufactured in a controlled environment and is intended for applications where ready-to-use agar strips do not meet the user′s requirements. Open the wrapper at 1/3 by pealing back the plastic seal at the rounded side of the wrapper. Remove the strip from the wrapper. Fill the strip evenly with 10 mL of a pre-prepared agar medium by applying aseptic techniques and allow it to solidify. Replace the filled strip into the original wrapper and close the wrapper with the included cover slide until use. The strips are designed to be used with RCS® Air Samplers.

Application

Agar Strips are used for monitoring of ambient air and compressed gases with the RCS® Air Samplers.

Legal Information

HYCON is a registered trademark of Merck KGaA, Darmstadt, Germany
RCS is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

For easy and flexible microbiological monitoring of surfaces in the food & beverage industry.

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service