Skip to Content
Merck
All Photos(1)

Key Documents

1.44024

Millipore

Hycon® Contact Slides

Empty slides, stand alone, sterile, pack of 20 units, suitable for surface monitoring

Synonym(s):

Hycon® Contact Slides Blank Slide Kit

Sign Into View Organizational & Contract Pricing

Select a Size

20 UNITS
€55.10

€55.10


Estimated to ship on15 April 2025Details


Request more information

Select a Size

Change View
20 UNITS
€55.10

About This Item

UNSPSC Code:
41106200
NACRES:
NA.74

€55.10


Estimated to ship on15 April 2025Details


Request more information

Quality Level

sterility

sterile

form

plate (ready-to-use)

feature

ready-to-use

packaging

pack of 20 units

manufacturer/tradename

HYCON®

application(s)

cosmetics
food and beverages
pharmaceutical

surface monitoring

compatibility

stand alone

storage temp.

15-25°C

suitability

bacteria
fungi
nonselective for

General description

Blank Strip Kit, Contact Slides are developed for easy and flexible microbiological monitoring of flat and curved surfaces. The unique design makes it a universal tool for aseptic environments and isolators. The Blank Strip Kit consists of 20 empty, sterile, and sealed slides, with a contact area of 25 cm2 It is intended for applications where ready-to-use contact slides do not meet the operator′s requirements.

Application

Blank slides are used for special applications in microbial surface monitoring.

Legal Information

HYCON is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service