Skip to Content
Merck

1.37035

SAFC

Acetic acid 1 mol/L

EMPROVE® EXPERT

Pharma Manufacturing

Synonym(s):

Acetic acid solution

Sign Into View Organizational & Contract Pricing

Select a Size

2.5 L
€50.00
25 L
€472.00
950 L
€5,657.00

€50.00


Estimated to ship on02 June 2026Details


Request a Bulk OrderRequest a Sample

Select a Size

Change View
2.5 L
€50.00
25 L
€472.00
950 L
€5,657.00

About This Item

Linear Formula:
CH3CO2H
CAS Number:
Molecular Weight:
60.05
Beilstein:
506007
MDL number:
UNSPSC Code:
12352301

€50.00


Estimated to ship on02 June 2026Details


Request a Bulk OrderRequest a Sample

Quality Level

product line

EMPROVE® EXPERT

form

liquid

pH

3 (20 °C in H2O)

density

1.00 g/cm3 at 20 °C

storage temp.

15-25°C

SMILES string

CC(O)=O

InChI

1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)

InChI key

QTBSBXVTEAMEQO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service