Skip to Content
Merck
All Photos(1)

Key Documents

1.30102

Millipore

HY-LiTE® Liquid Test Pens for Total ATP

Test pens for detection of Total ATP in liquid samples using HY-LiTE® 2/2A luminometers

Synonym(s):

Liquid ATP pens, Liquid ATP swabs, Liquid ATP tests, Liquid ATP samplers

Sign Into View Organizational & Contract Pricing

Select a Size

50 TESTS
€308.00

€308.00


Available to ship on04 April 2025Details


Request more information

Select a Size

Change View
50 TESTS
€308.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.74

€308.00


Available to ship on04 April 2025Details


Request more information

Quality Level

product line

HY-LiTE®

packaging

pkg of 50 tests

manufacturer/tradename

HY-LiTE®

application(s)

food and beverages
water monitoring

compatibility

for use with HY-LITE® luminometer

detection method

bioluminescent

storage temp.

2-8°C

General description

The HY-LiTE® Liquid Test Pens for Total ATP are ready prepared pens to measure Total ATP (adenosine triphosphate) in liquid samples using the HY-LiTE® 2/2A luminometer. ATP is detected by reaction with a luciferin/luciferase reagent in buffered solution.

Application

The HY-LiTE® Liquid Test Pens for Total ATP are used to test the cleanliness/hygiene of liquids (CIP rinse water) in food & beverage industry, or to test biomass in water treatment application.

Suitability

For detection of Total ATP in liquid samples.

Linkage

SKU 1.30102.0022 replaced by 1.30102.0021

Analysis Note

packaging: conforms
Appearance: passes test
Performance test: conforms

Legal Information

HY-LITE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Related Content

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service