Skip to Content
Merck
All Photos(1)

Key Documents

1.30101

Millipore

HY-LiTE® Surface ATP Test Kit

Test pens and hygiene swabs for use with HY-LiTE® 2/2A luminometers

Synonym(s):

Surface ATP pens, Surface ATP swabs, Surface ATP tests, Surface ATP samplers

Sign Into View Organizational & Contract Pricing

Select a Size

100 TESTS
€674.00

€674.00


Estimated to ship on11 April 2025Details


Request more information

Select a Size

Change View
100 TESTS
€674.00

About This Item

UNSPSC Code:
41112601
NACRES:
NA.74

€674.00


Estimated to ship on11 April 2025Details


Request more information

size

100 reactions

Quality Level

product line

HY-LiTE®

quality

Test pens and hygiene swabs for use with HY-LiTE® 2/2A luminometers

packaging

pkg of 100 tests (100 pens & 100 hygiene swabs)

manufacturer/tradename

HY-LiTE®

storage condition

protect from light

application(s)

food and beverages
personnel monitoring
surface monitoring

compatibility

for use with HY-LITE® luminometer

detection method

bioluminescent

storage temp.

2-8°C

General description

The HY-LiTE® Surface ATP Test Kit contains ready-prepared pens in cuvette test format for use with HY-LiTE® 2/ luminometers. The kit contains test pens and hygiene swabs that are primarily designed for the examination of post-production cleaning/sanitation effectiveness on product contact surfaces. Adenosine triphosphate (ATP) is detected by reaction with a luciferin/luciferase reagent in a buffered solution. The test kit is designed to indicate the presence of residues of biological origin, as well as bacteria, yeast and molds, including viable, non-culturable bacteria, anaerobes, and other microorganisms which are usually not detected by traditional growth-based methods.

Application

HY-LiTE® Surface ATP Test Kit is suitable for measuring ATP on surfaces after cleaning to monitor quality of sanitation processes.

Suitability

For surface hygiene monitoring

Linkage

SKU 1.30101.0022 replaced by 1.30101.0021

Analysis Note

packaging: conforms
Appearance: passes test
Performance test: conforms

Legal Information

HY-LITE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Related Content

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service