125185M
Eshmuno® CMX Resin
MiniChrom Column Eshmuno® CMX, volume 1 mL, prepacked column
About This Item
Recommended Products
ligand
(Carboxyethyl with Alkyl-Functional Groups)
material
polypropylene housing
sterility
non-sterile
product line
Eshmuno®
form
prepacked column
parameter
≤8 bar pressure
300 cm/hr max. flow rate
volume
1 mL
matrix active group
Polyvinyl Ether
mean particle size
50 μm
capacity
~60 mL, resin binding capacity
application(s)
ADC
mAb
recombinant protein
storage temp.
15-25°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Features and Benefits
- Intensify processes and decrease costs by reducing the number of chromatographic steps and buffer consumption
- Improve performance with higher recovery rates, high selectivity and superior dynamic binding capacity
- Improve the user experience with a broad operational window, simplified process development and easy column packing due to a rigid base bead
Packaging
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Learn more on mAb downstream processing, more specifically polishing chromatography and the relevant associated products.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service