Recommended Products
ligand
(Sulfoisobutyl)
Quality Level
sterility
sterile (Caustic Stable)
form
resin
specific analyte(s)
MAbs
parameter
500 cm/hr flow rate
8 bar max. pressure
matrix active group
polymer, polyvinyl ether
mean particle size
50 μm
pH
7 ( in H2O)
capacity
120 mg binding capacity (lysozyme/ml of resin)
transition temp
flash point 35 °C
density
1 g/cm3
application(s)
ADC
mAb
recombinant protein
separation technique
strong cation exchange
storage temp.
2-30°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Features and Benefits
- High aggregate removal efficiency combined with high dynamic binding capacity
- High loading capacity combined with high purity at lower elution volumes.
- Wide pH and conductivity operating window
Packaging
- 1.20083.0100: Eshmuno® CPX Resin 100ml
- 1.20083.0010: Eshmuno® CPX Resin 10ml
- 1.20083.0500: Eshmuno® CPX Resin 500ml
- 1.20083.5000: Eshmuno® CPX Resin 5L
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
95.0 °F
Flash Point(C)
35 °C
What is the Emprove® Program?
The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:
Free of charge - Sign in to download
Free of charge - Sign in to download
Available by subscription or for a fee
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service