Skip to Content
Merck
All Photos(2)

Key Documents

1.25170

Millipore

MiniChrom Column Eshmuno® CP-FT, 0.2ml

column I.D. × L 5 mm × 10 mm

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€153.00

€153.00


Estimated to ship on28 March 2025Details



Select a Size

Change View
1 EA
€153.00

About This Item

UNSPSC Code:
41115711

€153.00


Estimated to ship on28 March 2025Details


ligand

(Sulfoisobutyl)

material

polypropylene housing

product line

Eshmuno®

parameter

≤8 bar pressure
400 cm/hr max. flow rate

column I.D. × L

5 mm × 10 mm

volume

0.2 mL

matrix active group

Polyvinyl Ether

mean particle size

50 μm

capacity

~70 mL, resin binding capacity

application(s)

mAb

storage temp.

15-25°C

General description

5 x 10mm

Features and Benefits

Eshmuno® CP-FT chromatography resin enables:

  • Superior flow-through removal of mAb aggregates
  • High product recoveries at high mass loadings
  • Significant reduction in resin and buffer volume
  • Smaller manufacturing footprint (smaller columns, buffer tanks, etc.)
  • Low salt process conditions eliminate the need for dilution before subsequent ion exchange steps
  • Significant reduction in processing volumes improves virus filtration and ultrafiltration processing economics
  • Rigid base bead enables higher flow rates and easier column packing
  • Strong chemical stability
  • Compatibility with 1 % (v/v) aqueous benzyl alcohol containing 150 mM NaCl and storage solutions 0.1 M NaOH

Packaging

Plastic bag(s)

Legal Information

Eshmuno is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Learn more on mAb downstream processing, more specifically polishing chromatography and the relevant associated products.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service