Skip to Content
Merck
All Photos(1)

Key Documents

12-405

Sigma-Aldrich

Histone H4 Peptide

biotin conjugate

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€409.00

€409.00


Estimated to ship on21 March 2025Details



Select a Size

Change View
100 μG
€409.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

€409.00


Estimated to ship on21 March 2025Details


Product Name

Histone H4 Peptide, biotin conjugate, This gene is intronless & encodes a member of the histone H4 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element.

Quality Level

manufacturer/tradename

Upstate®

technique(s)

affinity precipitation assay: suitable
immunoprecipitation (IP): suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Quality

Routinely evaluated as a substrate for histone-modifying enzymes.

Physical form

Lyophilized powder

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Role of 14-3-3? in poor prognosis and in radiation and drug resistance of human pancreatic cancers.
Zhaomin Li,Zizheng Dong,David Myer,Michele Yip-Schneider,Jianguo Liu,Ping Cui et al.
BMC Cancer null
Expression of cyclooxygenase-2, P-glycoprotein and multi-drug resistance-associated protein in canine transitional cell carcinoma.
J-Y Lee,S Tanabe,H Shimohira,Y Kobayashi,T Oomachi,S Azuma,K Ogihara,H Inokuma
Research in Veterinary Science null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service