Skip to Content
Merck
All Photos(1)

Key Documents

124008

Sigma-Aldrich

Akt Inhibitor II

The Akt Inhibitor II controls the biological activity of Akt. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Akt Inhibitor II, SH-5

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€409.00

€409.00


Estimated to ship on02 April 2026Details



Select a Size

Change View
1 MG
€409.00

About This Item

Empirical Formula (Hill Notation):
C29H59O10P
Molecular Weight:
598.75
UNSPSC Code:
12352200
NACRES:
NA.77

€409.00


Estimated to ship on02 April 2026Details


Quality Level

Assay

≥98% (NMR)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell permeable, reversible, and substrate competitive phosphatidylinositol analog that inhibits the activation of Akt and selected downstream substrates without affecting the phosphorylation of PDK-1 and other downstream kinases. Decreases phosphorylation of Akt without affecting the total Akt level. Reported to be a potent inducer of apoptosis and to selectively kill a variety of cancer cell lines expressing high levels of active Akt. When used at a concentration of 10 µM, it is reported to activate p38α/SAPK2α MAP kinase. At 10 µM it will also inhibit ~50% of PKA activity.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Akt
Product does not compete with ATP.
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Gills, J.J., et al. 2007. J. Biol. Chem.in press.
Kozikowski, A.P., et al. 2003. J. Am. Chem. Soc.125, 1144.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rakan Radi et al.
Antioxidants (Basel, Switzerland), 11(5) (2022-05-29)
The development of targeted therapies (BRAF/MEK inhibitors) and immunotherapy have had a major impact on the treatment of melanoma. However, the majority of patients with advanced melanomas succumb to their disease. The mechanisms of resistance to both targeted therapies and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service