Skip to Content
Merck
All Photos(1)

Key Documents

123871

Sigma-Aldrich

SC79

≥98% (sum of two isomers, HPLC), solid, Akt activator, Calbiochem®

Synonym(s):

Akt Activator II, SC79, Ethyl-2-amino-6-chloro-4-(1-cyano-2-ethoxy-2-oxoethyl)-4H-chromene-3-carboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€242.00

€242.00


Estimated to ship on14 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 MG
€242.00

About This Item

Empirical Formula (Hill Notation):
C17H17ClN2O5
CAS Number:
Molecular Weight:
364.78
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€242.00


Estimated to ship on14 April 2025Details


Request a Bulk Order

Product Name

Akt Activator II, SC79, Akt Activator II, SC79, CAS 305834-79-1, is a cell-permeable compound that interacts with Akt PH domain PIP3 binding pocket and makes it more susceptible to phosphorylation by upstream Kinases.

Quality Level

Assay

≥98% (sum of two isomers, HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

Clc1cc2c(cc1)OC(=C(C2C(C(=O)OCC)C#N)C(=O)OCC)N

InChI

1S/C17H17ClN2O5/c1-3-23-16(21)11(8-19)13-10-7-9(18)5-6-12(10)25-15(20)14(13)17(22)24-4-2/h5-7,11,13H,3-4,20H2,1-2H3

InChI key

DXVKFBGVVRSOLI-UHFFFAOYSA-N

General description

A cell-permeable and blood-brain barrier permeant bromo-to-chloro substituted HA14-1 analog that renders Akt in a conformation susceptible to phosphorylation by upstream kinases via specific interaction with Akt pleckstrin homology (PH) domain PtdIns(3,4,5)P3- (PIP3) binding pocket. Shown to enhance both basal and receptor-mediated Akt phosphorylation (Thr308 and Ser473) in a time- and dose-dependent manner (2 to 4 µg/ml) with concomitant inhibition Akt membrane translocation in various cell cultures. Efficiently reduces glutamate-induced neurotoxicity both in primary neuron cultures (EC50 = 4 µg/ml) in vitro and in a murine MCAO (middle cerebral artery occlusion) model (40 mg/kg, i.p.) in vivo.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Pleckstrin homology (PH) domain of Akt

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and immediately freeze (-70°C). Stock solutions are stable for up to 2 weeks at -70°C.
Use only fresh DMSO for reconstitution.

Other Notes

Jo. H., et al. 2012. Proc. Natl. Acad. Sci. USA109, 10581.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service