Skip to Content
Merck

1.20084

Millipore

Eshmuno® CPS Resin

Synonym(s):

Eshmuno® CPS

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
€104.00
100 ML
€844.00
500 ML
€2,975.00
5000 ML
Inquire

€104.00


Estimated to ship on07 April 2025Details



Select a Size

Change View
10 ML
€104.00
100 ML
€844.00
500 ML
€2,975.00
5000 ML
Inquire

About This Item

UNSPSC Code:
41115711

€104.00


Estimated to ship on07 April 2025Details


Quality Level

ligand

(Sulfoisobutyl)

sterility

sterile (Caustic Stable)

product line

Eshmuno®

form

resin

parameter

500 cm/hr flow rate
8 bar max. pressure

matrix active group

polymer, polyvinyl ether

mean particle size

50 μm

capacity

160 mg binding capacity (lysozyme/ml of resin)

transition temp

flash point 35 °C

density

1 g/cm3

application(s)

recombinant protein

separation technique

cation exchange
weak cation exchange

storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

General description

High capacity and salt tolerant cation exchange resin for recombinant protein purification

Features and Benefits

Eshmuno® CPS chromatography resin enables:

  • Outstanding binding capacities at elevated conductivity levels (≥ 10 mS/cm)
  • Excellent separation efficiencies at high protein loadings
  • Ease of implementation and selection of process parameters through strong cation exchanger chemistry without hydrophobic groups
  • Established Fab fragments, recombinant human growth factor, and recombinant proteins purification protocols
  • High flow rate versus pressure-flow behavior
  • Excellent removal of impurities
  • Robust and safe packing procedures
  • Strong chemical stability
  • Compatibility with 1 % (v/v) aqueous benzyl alcohol containing 150 mM NaCl and storage solutions 0.1 M NaOH

Packaging

  • 1.20084.0100: Eshmuno® CPS Resin 100ml
  • 1.20084.0010: Eshmuno® CPS Resin 10ml
  • 1.20084.0500: Eshmuno® CPS Resin 500ml
  • 1.20084.5000: Eshmuno® CPS Resin 5L

Plastic bottle

Legal Information

Eshmuno is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

95.0 °F

Flash Point(C)

35 °C


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service