Skip to Content
Merck
All Photos(1)

Key Documents

1.19874

Supelco

Disposable PTFE liner for LiChrolut®vacuum manifold

Sign Into View Organizational & Contract Pricing

Select a Size

100 UNITS
€223.00

€223.00


Estimated to ship on24 March 2025Details


Request a Bulk OrderRequest a Sample

Select a Size

Change View
100 UNITS
€223.00

About This Item

UNSPSC Code:
41115711
NACRES:
NB.21

€223.00


Estimated to ship on24 March 2025Details


Request a Bulk OrderRequest a Sample

technique(s)

solid phase extraction (SPE): suitable

Quality Level

storage temp.

no temp limit

Legal Information

LICHROLUT is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Explore our selection of high-quality LiChrolut® SPE products for efficient and reliable sample preparation before chromatographic analysis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service