Skip to Content
Merck
All Photos(1)

Key Documents

1.19852

Supelco

LiChrolut® Drying unit

suitable for sample preparation

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
€1,550.00

€1,550.00


Estimated to ship on28 March 2025Details


Request a Bulk OrderRequest a Sample

Select a Size

Change View
1 UNIT
€1,550.00

About This Item

UNSPSC Code:
41115711
NACRES:
NB.21

€1,550.00


Estimated to ship on28 March 2025Details


Request a Bulk OrderRequest a Sample

technique(s)

solid phase extraction (SPE): suitable

Quality Level

application(s)

sample preparation

storage temp.

no temp limit

General description

All the individual steps associated with solid-phase extraction can be carried out using the LiChrolut extraction unit rapidly and reliability.This transparent and vacuum – suitable unit made of glass can be used to prepare up to 12 samples simultaneously.
for sample preparation

Legal Information

LICHROLUT is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Explore our selection of high-quality LiChrolut® SPE products for efficient and reliable sample preparation before chromatographic analysis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service