Skip to Content
Merck
All Photos(2)

Key Documents

1.19203

Supelco

Silver chloride 99+

for analysis 99+ EMSURE®

Synonym(s):

Silver chloride 99+

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€448.00
100 G
€1,350.00
1 KG
€8,970.00

€448.00


Estimated to ship on14 May 2025Details


Request a Bulk Order

Select a Size

Change View
25 G
€448.00
100 G
€1,350.00
1 KG
€8,970.00

About This Item

Empirical Formula (Hill Notation):
AgCl
CAS Number:
Molecular Weight:
143.32
MDL number:
UNSPSC Code:
12352302
EC Index Number:
232-033-3
NACRES:
NA.21
form:
powder
solubility:
0.00188 g/L

€448.00


Estimated to ship on14 May 2025Details


Request a Bulk Order

Quality Level

product line

EMSURE®

Assay

99.4-100.0% Ag basis

form

powder

potency

>5110 mg/kg LD50, oral (Rat)

mp

455 °C

solubility

0.00188 g/L

density

5.56 g/cm3 at 20 °C

cation traces

Ag: 74.9-75.3%
Cd: ≤0.002%
Cu: ≤0.001%
Fe: ≤0.001%
Pb: ≤0.002%

storage temp.

15-25°C

SMILES string

[Ag]Cl

InChI

1S/Ag.ClH/h;1H/q+1;/p-1

InChI key

HKZLPVFGJNLROG-UHFFFAOYSA-M

Related Categories

Analysis Note

Identity (elemental analysis): passes test
Assay (calculated of Ag): 99.4 - 100.0 %
Ag (Silver): 74.9 - 75.3 %
Cu (Copper): ≤ 0.001 %
Pb (Lead): ≤ 0.002 %
Fe (Iron): ≤ 0.001 %
Cd (Cadmium): ≤ 0.002 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Met. Corr. 1

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service