Key Documents
1.17247
Supelco
RQcheck set
Select a Size
1 SET
€57.90
€57.90
Available to ship on31 March 2025Details
Request a Bulk OrderRequest more information
Select a Size
Change View
1 SET
€57.90
About This Item
UNSPSC Code:
41116116
NACRES:
NB.35
€57.90
Available to ship on31 March 2025Details
Request a Bulk OrderRequest more information
compatibility
RQflex® 20
Quality Level
detection method
reflectometric
General description
The RQcheck set intended for use with the Reflectoquant® RQflex® 20 reflectometer. It allows performing the selectable quality assurance procedure "RQcheck" to ensure nominal functionality of the instrument. The set contains a grey check strip and an corresponding barcode strip.
Packaging
Plastic bag
Legal Information
REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
RQFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany