Skip to Content
Merck
All Photos(1)

Documents

1.16957

Supelco

Check Set

for use with RQFLEX® 10, RQFLEX® 10 plus

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115316
NACRES:
NB.35

Quality Level

compatibility

for use with RQFLEX 10, RQFLEX 10 plus

detection method

reflectometric

storage temp.

15-25°C

General description

RQcheck RQflex® 10, RQflex® plus Reflectoquant®
We offer you useful accessories. The check set can be used for RQflex® 10 and the RQflex® 10 plus reflectometer. The set contains a grey check strip and two barcode strips.

Linkage

Replaces: 1.16957.0007

Legal Information

REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
RQFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service