Skip to Content
Merck
All Photos(2)

Key Documents

1.14438

Supelco

Iron Test Kit, colorimetric

0.2-2.5 mg/L (Fe), MQuant®

Synonym(s):

Iron Visual Test Kit

Sign Into View Organizational & Contract Pricing

Select a Size

500 TESTS
€185.00

€185.00


Estimated to ship on03 April 2025Details


Request a Bulk OrderRequest more information

Select a Size

Change View
500 TESTS
€185.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

€185.00


Estimated to ship on03 April 2025Details


Request a Bulk OrderRequest more information

Product Name

Iron Test, colorimetric, 0.2-2.5 mg/L (Fe), for use with MCOLORTEST®

specific analyte(s)

iron

Quality Level

measuring range

0.2-2.5 mg/L (Fe)

compatibility

for use with MCOLORTEST®

detection method

colorimetric

storage temp.

15-25°C

General description

Method: colorimetric with color card and comparator block with short test tubes 0.2 - 0.4 - 0.6 - 0.8 - 1.0 - 1.3 - 1.6 - 2.0 - 2.5 mg/l Fe MQuant®
The MQuant® Iron test (formally MColortest®) measures bivalent and trivalent iron in its dissolved form as well as fresh colloidal iron(III) hydroxide in
  • Groundwater and surface water, seawater
  • Drinking water and mineral water
  • Waters from aquaculture
  • Boiler and boiler feed water, cooling water
  • Industrial water
  • Wastewater and percolating water
  • Food after appropriate sample pretreatment

Method
All iron ions are reduced to iron(II) ions. In a thioglycolate-buffered medium these react with a triazine derivative to form a red-violet complex. The iron concentration is measured semiquantitatively by visual comparison of the color of the measurement solution with the color fields of a color card. The unique brilliance of the print and their fine color graduation allow precise analyses, even at the lower end of the concentration range.
Using the refill pack 1.18458 containing all the reagents, the original test kit can be used for a very long time, making it a particularly economical and ecological system. 

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service