Skip to Content
Merck
All Photos(1)

Key Documents

1.18458

Supelco

Iron Test Refill pack

for use with MCOLORTEST®

Synonym(s):

Iron Visual Test Kit Refills

Sign Into View Organizational & Contract Pricing

Select a Size

500 TESTS
€175.00

€175.00


Estimated to ship on09 April 2025Details


Request a Bulk OrderRequest more information

Select a Size

Change View
500 TESTS
€175.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

€175.00


Estimated to ship on09 April 2025Details


Request a Bulk OrderRequest more information

feature

refill

Quality Level

specific analyte(s)

iron

compatibility

for use with MCOLORTEST®

detection method

colorimetric

storage temp.

15-25°C

General description

When the reagents of a MQuant® test (formally MColortest®) are used up, the kit can be replenished by ordering the appropriate refill pack. Refill packs are available for a range of test kits, making it a particularly economical and ecological system.
This refill pack contains all reagents for the MQuant® test kits 1.14759, 1.14438, and 1.14403 for the colometric evaluation of iron.

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service