Skip to Content
Merck
All Photos(1)

Key Documents

1.11748

Supelco

Phenol red sodium salt

pH indicator, ACS

Synonym(s):

Phenol red sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€64.40

€64.40


Estimated to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
5 G
€64.40

About This Item

Empirical Formula (Hill Notation):
C19H13NaO5S
CAS Number:
Molecular Weight:
376.36
MDL number:
UNSPSC Code:
12171502
EC Index Number:
252-057-8
NACRES:
NA.21

€64.40


Estimated to ship on31 March 2025Details


Request a Bulk Order

Product Name

Phenol red sodium salt, indicator ACS

grade

ACS reagent

Quality Level

loss

≤8.0% loss on drying, 110°C

visual transition interval

1.2-3.0, brown-orange to yellow(1st transition range)
6.5-8, brownish-yellow to red-violet(2nd transition range)

mp

285 °C

storage temp.

2-30°C

SMILES string

[Na+].[S]1(=O)(=O)OC(c4c1cccc4)(c3ccc(cc3)[O-])c2ccc(cc2)O

InChI

1S/C19H14O5S.Na/c20-15-9-5-13(6-10-15)19(14-7-11-16(21)12-8-14)17-3-1-2-4-18(17)25(22,23)24-19;/h1-12,20-21H;/q;+1/p-1

InChI key

HKHYOKBQJILTEI-UHFFFAOYSA-M

Analysis Note

1st transition range: pH 1.2 - pH 3.0brownish orange - yellow
2nd transition range: pH 6.5 - pH 8.0brownish yellow - red violet
Appearance of solution: passes test
Loss on drying (110 °C): ≤ 8.0 %
Transition range (according to ACS): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service