Skip to Content
Merck
All Photos(1)

Documents

1.59375

Supelco

Phenol red

pH indicator, Reag. Ph Eur

Synonym(s):

Phenol red

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H4OH)2C7H4SO3
CAS Number:
Molecular Weight:
354.38
MDL number:
UNSPSC Code:
12171502
EC Index Number:
205-609-7
NACRES:
NA.21

product name

Phenol red, (phenolsulfonphthalein) Reag. Ph Eur

Agency

reag. Ph. Eur.

Quality Level

Assay

98.0-102.0% (bromatometric, calculated on dried substance)

form

solid

loss

≤1.0% loss on drying, 105°C

mp

>300 °C

bulk density

200‑300 kg/m3

storage temp.

no temp limit

InChI

1S/C19H14O5S/c20-15-9-5-13(6-10-15)19(14-7-11-16(21)12-8-14)17-3-1-2-4-18(17)25(22,23)24-19/h1-12,20-21H

InChI key

BELBBZDIHDAJOR-UHFFFAOYSA-N

Related Categories

Application

  • Antitussive, Expectorant and Antipyretic Effect of the Ethanolic Extract of the Leaves of Momordica charantia L.: This study investigates the application of phenol red in assessing the bioactivities of plant extracts, demonstrating the versatility of phenol red in chemical assays (Ribeiro et al., 2024).
  • Chemical Analysis by LC-MS of Cannabis sativa Root Samples from Northeast Brazil and Evaluation of Antitussive and Expectorant Activities.: Phenol red was used as part of the LC-MS methodology to analyze plant roots, highlighting its role in modern analytical techniques (Menezes et al., 2022).
  • Quantifying Sample Collection and Processing Impacts on Fiber-Based Tear Fluid Chemical Analysis.: Phenol red is utilized to quantify changes in tear fluid composition, underscoring its importance in clinical chemical analysis (Barmada & Shippy, 2020).
  • The In Vitro Activity of Essential Oils against Helicobacter Pylori Growth and Urease Activity.: This research uses phenol red in enzyme inhibition studies, contributing to the understanding of antimicrobial properties of essential oils (Korona-Glowniak et al., 2020).

Analysis Note

Assay (bromatometric, calculated on dried substance): 98.0 - 102.0 %
Identity (UV/VIS-Spectrum): passes test
Identity (Color transition): passes test
Identity (Bromination): passes test
Insoluble substances: ≤ 0.5
Sulfated ash: ≤ 0.2
Loss on Drying (105°C): ≤ 1.0

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service