Skip to Content
Merck
All Photos(1)

Key Documents

1.09275

Sigma-Aldrich

Shorr staining solution

for hormonal cytodiagnosis

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
€84.00

€84.00


Estimated to ship on27 March 2025Details


Request a Bulk Order

Select a Size

Change View
500 ML
€84.00

About This Item

UNSPSC Code:
41116113
NACRES:
NA.77

€84.00


Estimated to ship on27 March 2025Details


Request a Bulk Order

Quality Level

form

liquid

autoignition temp.

425 °C (ethanol)

IVD

for in vitro diagnostic use

technique(s)

microbe id | staining: suitable

transition temp

flash point 19 °C

density

0.91 g/cm3 at 20 °C

application(s)

clinical testing
diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

General description

Shorr staining solution - for hormonal cytodiagnosis, is a ready-to-use staining solution that is used for human-medical cell diagnosis and serves the cytological investigation of sample material of human origin, for example cervical smears. During the course of menstrual cycle, the sex hormones cause characteristic changes in the vaginal epithelium. The hormone situation can be evaluated with the aid of stained vaginal smears. The Shorr staining is used for hormonal cytodiagnosis only, however the differences between eosinophilia and cyanophilia are more distinct. The ratio of eosinophilic to cyanophilic cells casts light on the effects of the folliccular and the corpus luteum hormones. The number of eosinophilic cells increases under the influence of the follicular hormone, whilst that of the cyanophilic cells increases under the influence of corpus luteum hormone.
The 500 ml bottle will provide approx. 1000 stainings. The Shorr staining solution is an IVD registered product and CE certified, thus can be used for clinical diagnostic purposes. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Analysis Note

Suitability for microscopy (Vaginal smear): passes test
Nuclei: brownish red
Eosinophilic cytoplasm: red
Cyanophilic cytoplasm: blue-green

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

66.2 °F

Flash Point(C)

19 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service