Skip to Content
Merck
All Photos(1)

Key Documents

1.07284

Millipore

Gelatin Peptone for Biotechnology and Microbiology

Pancreatically Digested, Prime, for Biotechnology and Microbiology

Synonym(s):

Peptone from gelatin

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€326.00

€326.00


Estimated to ship on24 March 2025Details


Request more information

Select a Size

Change View
1 KG
€326.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106212
EC Index Number:
293-431-0
NACRES:
NA.74

€326.00


Estimated to ship on24 March 2025Details


Request more information

Product Name

Peptone from gelatin, pancreatic digested, suitable for microbiology

biological source

animal

Quality Level

Assay

15.5-17.0% amino-nitrogen basis (purity)

form

powder

pH

6-7 (20 °C, 20 g/L in H2O)

solubility

550 g/L

bulk density

420 kg/m3

application(s)

microbiology

storage temp.

15-25°C

Analysis Note

Appearance: Lightly yellow to lightly brownishyellow fine powder.
Appearance of solution (5 %; water): Clear and yellow to brownishyellow.
pH-value (5 %; water): 6.5 - 7.5
Amino nitrogen (as N): 2.2 - 3.2 %
Total nitrogen (N) (Kjeldahl): 15.5 - 17.0 %
Sulfated ash (800 °C): ≤ 15.0 %
Loss on Drying (105°C): ≤ 6.0 %
Ca (Calcium): ≤ 0.2 %
Suitability for microbiology: passes test
Identity (NIR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service