Skip to Content
Merck
All Photos(1)

Key Documents

1.07223

Supelco

1,10-Phenanthroline chloride monohydrate

GR for analysis and redox indicator [reagent for iron(II)] Reag. Ph Eur

Synonym(s):

1,10-Phenanthroline chloride monohydrate, o-Phenanthroline hydrochloride monohydrate

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€90.60
100 G
€793.00

€90.60


Estimated to ship on02 April 2025Details


Request a Bulk Order

Select a Size

Change View
10 G
€90.60
100 G
€793.00

About This Item

Empirical Formula (Hill Notation):
C12H9ClN2 · xH2O
CAS Number:
Molecular Weight:
216.67 (anhydrous basis)
MDL number:
UNSPSC Code:
12352107
EC Index Number:
223-325-1
NACRES:
NA.21

€90.60


Estimated to ship on02 April 2025Details


Request a Bulk Order

Agency

reag. Ph. Eur.

Quality Level

form

solid

potency

132 mg/kg LD50, oral (Rat)

mp

215-220 °C (decomposition)

bulk density

370 kg/m3

storage temp.

2-30°C

SMILES string

[Cl-].[n+H]1c2c3ncccc3ccc2ccc1.O

InChI

1S/C12H8N2.ClH.H2O/c1-3-9-5-6-10-4-2-8-14-12(10)11(9)13-7-1;;/h1-8H;1H;1H2

InChI key

NDLHUHRGAIHALB-UHFFFAOYSA-N

Analysis Note

Assay (alcalimetric, calculated on anhydrous substance): ≥ 99.5 %
Identity (IR-spectrum): passes test
Melting point: about 215 °C, with decomposition
Substances insoluble in water: passes test
Sulfated ash: ≤ 0.2 %
Water (according to Karl Fischer): 7.0 - 8.5 %
Suitability as indicator: passes test
Sensitivity test (at Iron II ions): passes test

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

The article describes the sample preparation and photometric determination of silver concentration in water samples using Spectroquant® photometers and test kits.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service