Skip to Content
Merck
All Photos(1)

Key Documents

1.03965

Supelco

Iron(II) sulfate heptahydrate

for analysis EMSURE® ACS,ISO,Reag. Ph Eur

Synonym(s):

Iron(II) sulfate heptahydrate, Iron

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€35.30
500 G
€76.50
1 KG
€118.00
5 KG
€412.00
25 KG
€2,830.00

€35.30


Estimated to ship on02 July 2025Details


Request a Bulk Order

Select a Size

Change View
100 G
€35.30
500 G
€76.50
1 KG
€118.00
5 KG
€412.00
25 KG
€2,830.00

About This Item

Empirical Formula (Hill Notation):
FeO4S · 7H2O
CAS Number:
Molecular Weight:
278.01
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-753-5
NACRES:
NA.21
Assay:
99.5-102.0% (manganometric)
grade:
ACS reagent
form:
solid

€35.30


Estimated to ship on02 July 2025Details


Request a Bulk Order

grade

ACS reagent

Quality Level

Agency

reag. ISO
reag. Ph. Eur.

product line

EMSURE®

Assay

99.5-102.0% (manganometric)

form

solid

potency

319 mg/kg LD50, oral (Rat)

impurities

≤0.01% Insoluble matter

pH

3-4 (20 °C, 50 g/L in H2O)

mp

>60 °C (Elimination of water of crystallisation)

density

1.89 g/cm3 at 20 °C

bulk density

600 kg/m3

anion traces

chloride (Cl-): ≤0.0005%
phosphate (PO43-): ≤0.001%

cation traces

As: ≤0.0002%
Ca: ≤0.005%
Co: ≤0.0025%
Cr: ≤0.0050%
Cu: ≤0.001%
Fe3+: ≤0.02%
K: ≤0.002%
Mg: ≤0.002%
Mn: ≤0.05%
N: ≤0.001%
Na: ≤0.02%
Ni: ≤0.0050%
Pb: ≤0.0005%
Zn: ≤0.0050%
heavy metals (as Pb): ≤0.005%

storage temp.

15-25°C

SMILES string

[Fe+2].[S](=O)(=O)([O-])[O-].O.O.O.O.O.O.O

InChI

1S/Fe.H2O4S.7H2O/c;1-5(2,3)4;;;;;;;/h;(H2,1,2,3,4);7*1H2/q+2;;;;;;;;/p-2

InChI key

SURQXAFEQWPFPV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Linkage

Replaces: FX0260-1

Analysis Note

Assay (manganometric): 99.5 - 102.0 %
Identity: passes test
Appearance of solution: passes test
Insoluble matter: ≤ 0.01 %
pH-value (5 %; water): 3.0 - 4.0
Chloride (Cl): ≤ 0.0005 %
Phosphate (PO₄): ≤ 0.001 %
Total nitrogen (N): ≤ 0.001 %
Heavy metals as Pb: ≤ 0.005 %
As (Arsenic): ≤ 0.0002 %
Ca (Calcium): ≤ 0.005 %
Cr (Chromium): ≤ 0.0050 %
Cu (Copper): ≤ 0.001 %
Fe III (Iron III): ≤ 0.02 %
K (Potassium): ≤ 0.002 %
Mg (Magnesium): ≤ 0.002 %
Mn (Manganese): ≤ 0.05 %
Na (Sodium): ≤ 0.02 %
Ni (Nickel): ≤ 0.0050 %
Pb (Lead): ≤ 0.0005 %
Zn (Zinc): ≤ 0.0050 %
Co (Cobalt): ≤ 0.0025 %
Substance not precipitated by ammonium hydroxide: ≤ 0.1 %
Corresponds to ACS,ISO,Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. To whom it may concern, We keep Iron(II) sulfate heptahydrate at room temperature over the years. I am wondering if there is a chance it can get oxidized?

    1 answer
    1. Iron(II) sulfate heptahydrate can oxidize over time, particularly when exposed to air. The recommended storage temperature is 15-25°C. The lot-specific shelf life is available on the Certificate of Analysis.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service