Skip to Content
Merck
All Photos(1)

Key Documents

1.06798

Supelco

4-Nitrophenol

pH indicator

Synonym(s):

4-Nitrophenol, p-Nitrophenol, 1-Hydroxy-4-nitrobenzol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5NO3
CAS Number:
Molecular Weight:
139.11
MDL number:
UNSPSC Code:
12352104
EC Index Number:
202-811-7
NACRES:
NA.21
Pricing and availability is not currently available.

Product Name

4-Nitrophenol, indicator

Quality Level

autoignition temp.

510 °C

potency

1024 mg/kg LD50, skin (Rat)

pH

4.4 (24 °C, 5 g/L in H2O)

bp

279 °C/1013 hPa (decomposition)

mp

110-114 °C

transition temp

flash point 169 °C

solubility

14.8 g/L

density

1.48 g/cm3 at 20 °C

bulk density

550 kg/m3

storage temp.

2-30°C

SMILES string

[N+](=O)([O-])c1ccc(cc1)O

InChI

1S/C6H5NO3/c8-6-3-1-5(2-4-6)7(9)10/h1-4,8H

InChI key

BTJIUGUIPKRLHP-UHFFFAOYSA-N

Application


  • Controlled Glycolysis of Poly(ethylene terephthalate): Research shows that using Antimony(III) Oxide for controlled glycolysis of PET under microwave irradiation results in well-defined oligomers, demonstrating its potential for effective polymer degradation (Mohammadi et al., 2023).



Analysis Note

Assay (DSC (differential scanning calorimetry)): ≥ 99.5 %
Transition range: pH 5.4 - pH 7.5colourless - yellow
Appearance of solution: passes test
Melting point (DSC (differential scanning calorimetry)): 110 - 114 °C

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - STOT RE 2 Oral

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

336.2 °F - closed cup

Flash Point(C)

169.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service