Skip to Content
Merck
All Photos(1)

Key Documents

P1852

Millipore

Pseudomonas agar

suitable for microbiology, NutriSelect® Plus

Synonym(s):

For Fluorescein

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€227.00

€227.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
500 G
€227.00

About This Item

UNSPSC Code:
41171609
NACRES:
NA.85

€227.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

sterility

non-sterile

Quality Level

product line

BioChemika

form

powder

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbe id | metabolite detection: suitable
microbiological culture: suitable

final pH

7.0±0.2 (25 °C)

application(s)

agriculture
environmental
food and beverages
water monitoring

microbiology

suitability

nonselective and differential for Pseudomonas spp.

Application

Recommended for the detection of fluorescein production by Pseudomonas.

Components

Ingredients (g/L)
Casein enzymic hydrolysate, 10.00
Proteose peptone, 10.00
Dipotassium phosphate, 1.50
Magnesium sulphate, 1.50
Agar, 15.00

Preparation Note

Suspend 38 g of Pseudomonas Agar (For Fluorescein) in 1000 ml of distilled water containing 10 ml of glycerol (Cat. No. 49767). Boil to dissolve the medium completely. Sterilize by autoclaving at 15 lbs. pressure (121°C) for 15 minutes.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Two simple media for the demonstration of pyocyanin and fluorescin.
King E O, et al.
Translational Research, 44(2), 301-307 (1954)
United States Pharmacopeia (1985)

Articles

An article concerning the detection, identification, differentiation, and cultivation of Pseudomonas species.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service