Skip to Content
Merck
All Photos(1)

Key Documents

1.06304

Supelco

Spectromelt® B 10

di-Sodium tetraborate, for X-ray fluorescence analysis

Synonym(s):

Borax

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€309.00
5 KG
€1,190.00

€309.00


Estimated to ship on03 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 KG
€309.00
5 KG
€1,190.00

About This Item

Empirical Formula (Hill Notation):
B4Na2O7
CAS Number:
Molecular Weight:
201.22
MDL number:
UNSPSC Code:
12352302
EC Index Number:
215-540-4
NACRES:
NA.21
form:
powder
solubility:
water: soluble 49.74 g/L at 20 °C (68 °F, Regulation (EC) No. 440/2008)

€309.00


Estimated to ship on03 April 2025Details


Request a Bulk Order

vapor pressure

7.3 hPa ( 1200 °C)

Quality Level

Assay

≥99.0% (acidimetric)

form

powder

potency

>2000 mg/kg LD50, skin (Rabbit)

pH

9.2 (20 °C, 25 g/L in H2O)

mp

742 °C

solubility

water: soluble 49.74 g/L at 20 °C (68 °F, Regulation (EC) No. 440/2008)

density

2.37 g/cm3 at 20 °C

bulk density

700 kg/m3

anion traces

chloride (Cl-): ≤50 ppm
phosphate (PO43-): ≤20 ppm
silicate (as SiO2): ≤100 ppm
sulfate (SO42-): ≤50 ppm

cation traces

Ag: ≤5 ppm
Al: ≤10 ppm
Ba: ≤5 ppm
Ca: ≤10 ppm
Cd: ≤5 ppm
Co: ≤5 ppm
Cr: ≤5 ppm
Cu: ≤5 ppm
Fe: ≤10 ppm
Ga: ≤5 ppm
K: ≤10 ppm
Mg: ≤5 ppm
Mn: ≤5 ppm
Ni: ≤5 ppm
Pb: ≤5 ppm
Sr: ≤5 ppm
Zn: ≤5 ppm

storage temp.

2-30°C

SMILES string

[Na+].[Na+].[B+3].[B+3].[B+3].[B+3].[O-2].[O-2].[O-2].[O-2].[O-2].[O-2].[O-2].O.O.O.O.O.O.O

InChI

1S/4B.2Na.7H2O.7O/h;;;;;;7*1H2;;;;;;;/q4*+3;2*+1;;;;;;;;7*-2

InChI key

HJIQVKXMQOLFFS-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): ≥ 99.0 %
Chloride (Cl): ≤ 50 ppm
Phosphate (PO₄): ≤ 20 ppm
Silicate (SiO₂): ≤ 100 ppm
Sulfate (SO₄): ≤ 50 ppm
Ag (Silver): ≤ 5 ppm
Al (Aluminium): ≤ 10 ppm
Ba (Barium): ≤ 5 ppm
Ca (Calcium): ≤ 10 ppm
Cd (Cadmium): ≤ 5 ppm
Co (Cobalt): ≤ 5 ppm
Cr (Chromium): ≤ 5 ppm
Cu (Copper): ≤ 5 ppm
Fe (Iron): ≤ 10 ppm
Ga (Gallium): ≤ 5 ppm
K (Potassium): ≤ 10 ppm
Mg (Magnesium): ≤ 5 ppm
Mn (Manganese): ≤ 5 ppm
Ni (Nickel): ≤ 5 ppm
Pb (Lead): ≤ 5 ppm
Sr (Strontium): ≤ 5 ppm
Zn (Zinc): ≤ 5 ppm
Loss on ignition (800 °C): ≤ 1.0 %
Bulk density: about 70

Legal Information

SPECTROMELT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service