Skip to Content
Merck

1.06303

SAFC

di-Sodium tetraborate decahydrate

EMPROVE® ESSENTIAL, Ph. Eur., BP, JP, NF

Pharma Manufacturing

Synonym(s):

di-Sodium tetraborate decahydrate, Borax, Sodium biborate decahydrate, Sodium borate decahydrate

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€72.00
3 X 1 KG
€250.00
25 KG
€1,169.00

€72.00


Estimated to ship on02 June 2025Details


Request a Bulk OrderRequest a Sample

Select a Size

Change View
1 KG
€72.00
3 X 1 KG
€250.00
25 KG
€1,169.00

About This Item

Empirical Formula (Hill Notation):
B4Na2O7 · 10H2O
CAS Number:
Molecular Weight:
381.37
MDL number:
UNSPSC Code:
12352302
EC Index Number:
603-411-9

€72.00


Estimated to ship on02 June 2025Details


Request a Bulk OrderRequest a Sample

Agency

BP
JP
NF
Ph. Eur.

Quality Level

vapor pressure

0.213 hPa ( 20 °C)

product line

EMPROVE® ESSENTIAL

Assay

99.0-103.0% (acidimetric)

form

solid

pH

9.2 (20 °C, 47 g/L in H2O)

mp

75 °C (Elimination of water of crystallisation)

solubility

49.74 g/L

density

1.72 g/cm3 at 20 °C

application(s)

liquid formulation
ophthalmics
pharmaceutical
semi-solid formulation

storage temp.

2-30°C

SMILES string

[Na+].[Na+].B([O-])(OB([O-])OB=O)OB=O.O.O.O.O.O.O.O.O.O.O

InChI

1S/B4O7.2Na.10H2O/c5-1-9-3(7)11-4(8)10-2-6;;;;;;;;;;;;/h;;;10*1H2/q-2;2*+1;;;;;;;;;;

InChI key

OJPFLODMGUZMKD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Sodium bromide is a mineral salt with diverse applications in pharmaceutical and cosmetic industry. It also has antimicrobial characteristics and is used as component of disinfectants.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service