Skip to Content
Merck
All Photos(1)

Documents

1.05326

Supelco

Lanthanum nitrate hexahydrate

for analysis

Synonym(s):

Lanthanum nitrate hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
La(NO3)3 · 6 H2O
CAS Number:
Molecular Weight:
433.01
MDL number:
UNSPSC Code:
12352302
EC Index Number:
233-238-0
NACRES:
NB.21

grade

for inorganic trace analysis

Quality Level

form

solid

potency

4500 mg/kg LD50, oral (Rat)

pH

3.4 (25 °C, 50 g/L in H2O)

bp

126 °C/1013 hPa (decomposition)

mp

69.9 °C

bulk density

1300 kg/m3

storage temp.

no temp limit

InChI

1S/La.3NO3.6H2O/c;3*2-1(3)4;;;;;;/h;;;;6*1H2/q+3;3*-1;;;;;;

InChI key

GJKFIJKSBFYMQK-UHFFFAOYSA-N

Application


  • Preparation of La-TiO2/Bentonite and Its Photodegradation Properties to Cyanide.: This study explores the application of Lanthanum nitrate hexahydrate in synthesizing La-TiO2/bentonite composite materials and their effectiveness in the photodegradation of cyanide, highlighting a potential application in pollutant removal techniques (Wang Q et al., 2016).

Analysis Note

Assay (complexometric): ≥ 96.0 %
Chloride (Cl): ≤ 0.005 %
Sulphate (SO₄): ≤ 0.005 %
Heavy metals (as Pb): ≤ 0.002 %
Ca (Calcium): ≤ 0.001 %
Ce (Cerium): ≤ 0.03 %
Fe (Iron): ≤ 0.002 %

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Ox. Sol. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service