Skip to Content
Merck
All Photos(1)

Key Documents

1.04973

Supelco

Potassium hexacyanoferrate(III)

for analysis EMSURE® ACS,Reag. Ph Eur

Synonym(s):

Potassium hexacyanoferrate(III), Potassium ferricyanide

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€99.30
250 G
€132.00
1 KG
€331.00

€99.30


Estimated to ship on24 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 G
€99.30
250 G
€132.00
1 KG
€331.00

About This Item

Linear Formula:
K3[Fe(CN)6]
CAS Number:
Molecular Weight:
329.24
MDL number:
UNSPSC Code:
12352302
EC Index Number:
237-323-3
NACRES:
NA.21
grade:
ACS reagent
form:
solid
solubility:
464 g/L

€99.30


Estimated to ship on24 March 2025Details


Request a Bulk Order

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

product line

EMSURE®

Assay

≥99.0% (iodometric)

form

solid

impurities

≤0.005% Insoluble matter

pH

6 (20 °C, 50 g/L in H2O)

solubility

464 g/L

density

1.85 g/cm3 at 20 °C

bulk density

900‑1000 kg/m3

anion traces

chloride (Cl-): ≤0.01%
hexacyanoferrate(II) ([Fe(CN)6]4-): ≤0.05%
sulfate (SO42-): ≤0.005%

cation traces

Pb: ≤0.002%

storage temp.

2-30°C

SMILES string

[Fe-3](C#N)(C#N)(C#N)(C#N)(C#N)C#N.[K+].[K+].[K+]

InChI

1S/6CN.Fe.3K/c6*1-2;;;;/q;;;;;;-3;3*+1

InChI key

MIMJFNVDBPUTPB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Silsesquioxane organofunctionalized with 4-amino-3-hydrazino-5-mercapto-1,2,4-triazole: Preparation and subsequent reaction with silver and potassium hexacyanoferrate(III) for detection of L-cysteine.: This study presents the preparation of a novel silsesquioxane composite functionalized with triazole, and its subsequent reaction with silver and potassium hexacyanoferrate(III). The resulting material is evaluated for its effectiveness in detecting L-cysteine, showcasing its potential in analytical chemistry applications (do Carmo et al., 2015).

  • Concerted derivatization and concentration method with dispersive liquid-liquid microextraction for liquid chromatographic analysis of 5-hydroxyindoles in human serum.: This research discusses a novel approach for the analysis of 5-hydroxyindoles in human serum using liquid chromatography. The method involves derivatization and concentration via dispersive liquid-liquid microextraction, enhancing the detection capabilities in complex biological matrices (Hayama et al., 2013).

  • Micro determination of cortisol and cortisone in umbilical cord blood by chemiluminescent high-performance liquid chromatography.: This paper focuses on the micro determination of cortisol and cortisone in umbilical cord blood using chemiluminescent HPLC. The study highlights the method′s sensitivity and precision for analyzing these important biomarkers (Hasegawa et al., 2010).

  • Comparative study of different cross-linking agents for the immobilization of functionalized carbon nanotubes within a chitosan film supported on a graphite-epoxy composite electrode.: This study evaluates various cross-linking agents for immobilizing functionalized carbon nanotubes within a chitosan film on a composite electrode. The findings have significant implications for enhancing electrode performance in electrochemical sensors (Pauliukaite et al., 2009).

  • Thin layer chromatography/fluorescence detection of 3,4-methylenedioxy-methamphetamine and related compounds.: This research explores the detection of MDMA and related compounds using thin-layer chromatography coupled with fluorescence detection. The method provides a reliable analytical technique for forensic applications (Kato et al., 2008).

Linkage

Replaces: PX1455; PX1455-1; PX1455-2; PX1455-4

Analysis Note

Assay (iodometric): ≥ 99.0 %
Insoluble matter: ≤ 0.005 %
Chloride (Cl): ≤ 0.01 %
Sulfate (SO₄): ≤ 0.005 %
Hexacyanoferrate (II) (Fe(CN)6)⁴⁻): ≤ 0.05 %
Pb (Lead): ≤ 0.002 %
Corresponds to ACS,Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2

Supplementary Hazards

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service