Skip to Content
Merck
All Photos(1)

Key Documents

1.05057

Supelco

Potassium disulfite

for analysis EMSURE®

Synonym(s):

Potassium disulfite, Potassium metabisulfite, Potassium pyrosulfite

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€49.40
1 KG
€76.10
2.5 KG
€154.00

€49.40


Estimated to ship on02 April 2025Details


Request a Bulk Order

Select a Size

Change View
500 G
€49.40
1 KG
€76.10
2.5 KG
€154.00

About This Item

Empirical Formula (Hill Notation):
K2O5S2
CAS Number:
Molecular Weight:
222.32
MDL number:
UNSPSC Code:
12352302
EC Index Number:
240-795-3
NACRES:
NA.21
form:
solid
solubility:
450 g/L

€49.40


Estimated to ship on02 April 2025Details


Request a Bulk Order

Quality Level

product line

EMSURE®

Assay

≥96% (iodometric)

form

solid

potency

2300 mg/kg LD50, oral (Rat)

impurities

≤0.005% In water insoluble matter

pH

3.0-4.5 (20 °C, 50 g/L in H2O)

mp

150 °C (decomposition)

solubility

450 g/L

bulk density

1000‑1300 kg/m3

anion traces

chloride (Cl-): ≤0.005%

cation traces

As: ≤0.0001%
Cu: ≤0.001%
Fe: ≤0.0005%
Pb: ≤0.001%
Zn: ≤0.001%

storage temp.

2-30°C

SMILES string

[K+].[K+].[S](=O)(=O)([S](=O)[O-])[O-]

InChI

1S/2K.H2O5S2/c;;1-6(2)7(3,4)5/h;;(H,1,2)(H,3,4,5)/q2*+1;/p-2

InChI key

RWPGFSMJFRPDDP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Analysis Note

Assay (iodometric): ≥ 96 %
In water insoluble matter: ≤ 0.005 %
Chloride (Cl): ≤ 0.005 %
As (Arsenic): ≤ 0.0001 %
Cu (Copper): ≤ 0.001 %
Fe (Iron): ≤ 0.0005 %
Pb (Lead): ≤ 0.001 %
Zn (Zinc): ≤ 0.001 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service