![Potassium disulfite analytical standard](/deepweb/assets/sigmaaldrich/product/structures/149/809/3c5b727d-e506-4214-889a-950c4d0873cb/640/3c5b727d-e506-4214-889a-950c4d0873cb.png)
Potassium disulfite
![Potassium disulfite analytical standard](/deepweb/assets/sigmaaldrich/product/structures/149/809/3c5b727d-e506-4214-889a-950c4d0873cb/640/3c5b727d-e506-4214-889a-950c4d0873cb.png)
Synonym(s):
Potassium metabisulfite, Potassium pyrosulfite, Potassium disulfite
Linear Formula:
K2S2O5
Products
Product Category
Brand
Formula Weight
pH Value
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Special Grade
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing