Skip to Content
Merck

1.05008

SAFC

2-Propanol

suitable for preparative chromatography, EMPROVE® ESSENTIAL, BP, USP

Pharma Manufacturing

Synonym(s):

sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€46.00
2.5 L
€74.00
25 L
€398.00

€46.00


Estimated to ship on02 April 2025Details


Request a Bulk OrderRequest a Sample

Select a Size

Change View
1 L
€46.00
2.5 L
€74.00
25 L
€398.00

About This Item

Linear Formula:
(CH3)2CHOH
CAS Number:
Molecular Weight:
60.10
Beilstein:
635639
MDL number:
UNSPSC Code:
12352104
EC Index Number:
200-661-7

€46.00


Estimated to ship on02 April 2025Details


Request a Bulk OrderRequest a Sample

Agency

BP
Ph. Eur.
USP

Quality Level

vapor density

2.1 (vs air)

vapor pressure

33 mmHg ( 20 °C)
43 hPa ( 20 °C)
44 mmHg ( 25 °C)

product line

EMPROVE® ESSENTIAL

Assay

≥99.8% (GC)

form

liquid

autoignition temp.

750 °F

expl. lim.

2.0-12.7 %, 93 °C

refractive index

n20/D 1.377 (lit.)

pH

7 (20 °C)

bp

82 °C (lit.)

mp

−89.5 °C (lit.)

transition temp

flash point 12 °C

density

0.785 g/mL at 25 °C (lit.)

application(s)

pharmaceutical

storage temp.

2-25°C

SMILES string

CC(C)O

InChI

1S/C3H8O/c1-3(2)4/h3-4H,1-2H3

InChI key

KFZMGEQAYNKOFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

2-Propanol is used for preparative chromatography.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12.0 °C - closed cup


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service