Skip to Content
Merck

1.04163

SAFC

Sodium chloride

granulated, EMPROVE® EXPERT, Ph. Eur., BP, ChP, JP, USP

Pharma Manufacturing

Synonym(s):

Halite, NaCl

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€55.00
10 KG
€175.00
3 X 1 KG
€220.00
25 KG
€329.00
500 KG
€3,598.00

€55.00


Estimated to ship on31 March 2025Details


Request a Bulk OrderRequest a Sample

Select a Size

Change View
1 KG
€55.00
10 KG
€175.00
3 X 1 KG
€220.00
25 KG
€329.00
500 KG
€3,598.00

About This Item

Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
Beilstein:
3534976
MDL number:
UNSPSC Code:
12352302

€55.00


Estimated to ship on31 March 2025Details


Request a Bulk OrderRequest a Sample

grade

GMO free
Halal
Kosher

Quality Level

Agency

BP
ChP
JP
Ph. Eur.
USP

product line

EMPROVE® EXPERT

form

granular

mp

801 °C (lit.)

application(s)

liquid formulation
parenterals
pharma/biopharma processes
pharmaceutical
solid formulation

storage temp.

2-30°C

SMILES string

[Na+].[Cl-]

InChI

1S/ClH.Na/h1H;/q;+1/p-1

InChI key

FAPWRFPIFSIZLT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Standard bulk materials often show a tendency for caking, clumping and dust formation which results in laborious weighing and manual de-caking processes with a negative impact on manufacturing efficiency. Due to its granulated form, Sodium chloride granulated is free-flowing and easy to handle. Its greatly reduced caking behavior facilitates processability, speeds up manufacturing processes and increases operator safety.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Sodium chloride is a mineral salt with broad range of applications in pharmaceutical formulations. It is typically used as an osmotic agent in liquid dosage forms e.g. for preparation of isotonic formulations, as bulking or coating agent for solid dosage forms, as well as for semi-solid dosage forms like creams.
Due to its low microbial and endotoxin limits, Sodium chloride granulated Emprove® Expert is especially suitable for high-risk applications.

Packaging

1.04163.0001 / 3x1kg / PE Bottle
1.04163.1000 / 1kg / PE Bottle
1.04163.9026 / 25kg / PE Bag in PE Drum
1.04163.9500 / 500kg / Big Bag

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Handling bulk powders at a large scale in the pharmaceutical, biopharmaceutical, and bioprocessing setting is challenging. Compared to powders, granulated materials experience far less caking, have better flowability, and are easier to handle, even after long-term storage.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service