Skip to Content
Merck
All Photos(2)

Key Documents

1.02711

Supelco

Copper(II) acetate monohydrate

greener alternative

for analysis EMSURE® ACS

Synonym(s):

Copper(II) acetate monohydrate, Cupric acetate

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€121.00
25 KG
€7,260.00

€121.00


Estimated to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
250 G
€121.00
25 KG
€7,260.00

About This Item

Linear Formula:
(CH3COO)2Cu · H2O
CAS Number:
Molecular Weight:
181.63 (anhydrous basis)
MDL number:
UNSPSC Code:
12352103
EC Index Number:
205-553-3
NACRES:
NA.21
grade:
ACS reagent
form:
solid
solubility:
72 g/L

€121.00


Estimated to ship on31 March 2025Details


Request a Bulk Order

grade

ACS reagent

Quality Level

product line

EMSURE®

Assay

99.0-102.0% (iodometric)

form

solid

potency

501 mg/kg LD50, oral (Rat)

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤0.01% Insoluble matter

pH

5.2-5.5 (20 °C, 20 g/L in H2O)

mp

115 °C

solubility

72 g/L

density

1.88 g/cm3 at 20 °C

bulk density

1100 kg/m3

anion traces

chloride (Cl-): ≤0.001%
sulfate (SO42-): ≤0.005%

cation traces

Ca: ≤0.005%
Fe: ≤0.002%
K: ≤0.01%
N: ≤0.01%
Na: ≤0.01%
Ni: ≤0.002%
Pb: ≤0.004%
Zn: ≤0.002%

greener alternative category

storage temp.

2-30°C

SMILES string

[Cu+2].[O-]C(=O)C.[O-]C(=O)C.O

InChI

1S/2C2H4O2.Cu.H2O/c2*1-2(3)4;;/h2*1H3,(H,3,4);;1H2/q;;+2;/p-2

InChI key

NWFNSTOSIVLCJA-UHFFFAOYSA-L

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product has been enhanced for catalysis. Click here for more information.

Application


  • Click-chemistry mediated synthesis of OTBN-1,2,3-Triazole derivatives exhibiting STK33 inhibition with diverse anti-cancer activities.: This study utilizes Copper(II) acetate monohydrate in the click-chemistry synthesis of OTBN-1,2,3-triazole derivatives, demonstrating their potential as inhibitors of serine/threonine kinase 33 (STK33), which could be significant for developing new anticancer treatments (Vala DP et al., 2024).

  • Synthesis and antifungal activity of slow-release pyridinylcarbonyl chitooligosaccharide copper complexes.: This research describes the synthesis of slow-release antifungal agents using Copper(II) acetate monohydrate to form copper complexes with pyridinylcarbonyl chitooligosaccharides, highlighting their efficacy against various fungal pathogens (Liu W et al., 2022).

  • Bis-triazolylchalcogenium-Functionalized Benzothiadiazole Derivatives as Light-up Sensors for DNA and BSA.: The paper reports on the development of bis-triazolylchalcogenium-functionalized benzothiadiazole derivatives synthesized using Copper(II) acetate monohydrate, which are used as fluorescent sensors for detecting DNA and bovine serum albumin (BSA) (Krüger R et al., 2021).

  • LED-driven photocatalysis of toluene, trichloroethylene and formaldehyde by cuprous oxide modified titanate nanotube arrays.: This study involves the use of Copper(II) acetate monohydrate in the synthesis of cuprous oxide-modified titanate nanotubes, enhancing the photocatalytic degradation of various organic pollutants under LED illumination (Chen CH et al., 2022).

Analysis Note

Assay (iodometric): 99.0 - 102.0 %
Insoluble matter: ≤ 0.01 %
Chloride (Cl): ≤ 0.001 %
Sulfate (SO₄): ≤ 0.005 %
Total nitrogen (N): ≤ 0.01 %
Ca (Calcium): ≤ 0.005 %
Fe (Iron): ≤ 0.002 %
K (Potassium): ≤ 0.01 %
Na (Sodium): ≤ 0.01 %
Ni (Nickel): ≤ 0.002 %
Pb (Lead): ≤ 0.004 %
Zn (Zinc): ≤ 0.002 %
Corresponds to ACS

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service