Skip to Content
Merck
All Photos(1)

Key Documents

1.02161

Supelco

Calcium sulfate dihydrate

precipitated for analysis EMSURE®

Synonym(s):

Calcium sulfate dihydrate

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€55.80
25 KG
€1,020.00

€55.80


Estimated to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
500 G
€55.80
25 KG
€1,020.00

About This Item

Empirical Formula (Hill Notation):
CaO4S · 2H2O
CAS Number:
Molecular Weight:
172.17
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-900-3
NACRES:
NA.21
form:
solid
solubility:
2 g/L

€55.80


Estimated to ship on31 March 2025Details


Request a Bulk Order

Quality Level

product line

EMSURE®

Assay

99.0-102.0% (complexometric)

form

solid

potency

>10000 mg/kg LD50, oral (Rat)

impurities

≤0.01% Substances insoluble in hydrochloric acid

pH

7.0 (20 °C, 50 g/L in H2O, slurry)

solubility

2 g/L

density

2.32 g/cm3 at 20 °C

bulk density

400‑600 kg/m3

anion traces

carbonate (CO32-): passes test
chloride (Cl-): ≤0.005%
nitrate (NO3-): passes test

cation traces

Fe: ≤0.001%
K: ≤0.005%
Mg: ≤0.01%
Na: ≤0.2%
Sr: ≤0.05%
heavy metals (as Pb): ≤0.002%

storage temp.

2-30°C

SMILES string

[Ca+2].[S](=O)(=O)([O-])[O-].O.O

InChI

1S/Ca.H2O4S.2H2O/c;1-5(2,3)4;;/h;(H2,1,2,3,4);2*1H2/q+2;;;/p-2

InChI key

PASHVRUKOFIRIK-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Acid leaching technology for post-consumer gypsum purification.: This research presents a novel method for purifying post-consumer gypsum, primarily composed of calcium sulfate dihydrate, through acid leaching, highlighting advancements in recycling and environmental management of gypsum waste (Castro-Diaz et al., 2023).

  • Gypsum-Related Impact on Antibiotic-Loaded Composite Based on Highly Porous Hydroxyapatite-Advantages and Disadvantages.: This study evaluates the influence of gypsum in antibiotic-loaded composites, offering significant findings on the biomedical applications of calcium sulfate dihydrate in drug delivery systems (Zalewska et al., 2023).

  • Organic solvent-free synthesis of calcium sulfate hemihydrate at room temperature.: Details a groundbreaking method for synthesizing calcium sulfate hemihydrate without organic solvents, contributing to greener and more sustainable chemical processes in industrial and laboratory settings (Reigl et al., 2024).

Analysis Note

Assay (complexometric): 99.0 - 102.0 %
Substances insoluble in hydrochloric acid: ≤ 0.01 %
Carbonate (CO₃): passes test
Chloride (Cl): ≤ 0.005 %
Nitrate (NO₃): passes test
Heavy metals (as Pb): ≤ 0.002 %
Fe (Iron): ≤ 0.001 %
K (Potassium): ≤ 0.005 %
Mg (Magnesium): ≤ 0.01 %
Na (Sodium): ≤ 0.2 %
Sr (Strontium): ≤ 0.05 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service