Skip to Content
Merck
All Photos(1)

Documents

1.00660

Supelco

Halogen lamp module

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115311
NACRES:
SG.01

compatibility

for use with Spectroquant® Pharo 100

storage temp.

2-30°C

General description

Pharo 100 Spectroquant®
The Halogen lamp module in the Spectroquant Pharo 100 can easily be replaced if necessary. Only use the lamp that is intended for use with your instrument. For other Photometers please choose the respective lamp.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service