Skip to Content
Merck
All Photos(1)

Key Documents

1.00099

Supelco

1-Amino-2-hydroxy-4-naphthalenesulfonic acid

for determination of phosphate

Synonym(s):

1-Amino-2-hydroxy-4-naphthalenesulfonic acid, 1-Amino-2-naphthol-4-sulfonic acid

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€123.00
100 G
€255.00

€123.00


Estimated to ship on11 June 2025Details


Request a Bulk Order

Select a Size

Change View
25 G
€123.00
100 G
€255.00

About This Item

Empirical Formula (Hill Notation):
C10H9NO4S
CAS Number:
Molecular Weight:
239.25
MDL number:
UNSPSC Code:
12352106
EC Index Number:
204-147-3
NACRES:
NB.21

€123.00


Estimated to ship on11 June 2025Details


Request a Bulk Order

Quality Level

form

solid

bulk density

280 kg/m3

storage temp.

2-30°C

SMILES string

[S](=O)(=O)(O)c1c2c(c(c(c1)O)N)cccc2

InChI

1S/C10H9NO4S/c11-10-7-4-2-1-3-6(7)9(5-8(10)12)16(13,14)15/h1-5,12H,11H2,(H,13,14,15)

InChI key

RXCMFQDTWCCLBL-UHFFFAOYSA-N

Application


  • 1-Amino-2-hydroxy-4-naphthalenesulfonic acid based Schiff bases or naphtho[1,2-d]oxazoles: selective synthesis and photophysical properties: This study explores the synthesis of Schiff bases and naphtho[1,2-d]oxazoles derived from 1-amino-2-hydroxy-4-naphthalenesulfonic acid, focusing on their photophysical properties. The research demonstrates potential applications in developing materials with specific optical properties, which could be significant in fields like organic electronics or photonics (Atahan & Durmus, 2015).

Linkage

Replaces: AX1035-1

Analysis Note

Assay (alkalimetric): ≥ 96 %
Identity (IR-spectrum): conforms
in sodium carbonate soluble components: conforms
Sulfated ash: ≤ 0.5 %
Loss on Drying (105 °C): ≤ 5.0 %
Performance test (for determination of phosphate): conforms

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service