100068
17-AAG
according to ISO 10272, A less toxic, potent, synthetic derivative of the ansamycin benzoquinone antibiotic Geldanamycin, Streptomyces hygroscopicus., GranuCult® prime
Synonym(s):
17-AAG, NSC-330507, 17-AG, 17-(Allylamino)-17-demethoxygeldanamycin, 17-Allylaminogeldanamycin
About This Item
Recommended Products
Agency
according to ISO 10272
Quality Level
product line
GranuCult® prime
Assay
≥98% (HPLC)
form
medium granules (dehydrated (DCM))
solid
manufacturer/tradename
Calbiochem®
GranuCult®
storage condition
OK to freeze
protect from light
technique(s)
microbiological culture: suitable
color
purple
solubility
DMSO: 10 mg/mL
methanol: 10 mg/mL
application(s)
food and beverages
microbiology
pathogen testing
shipped in
wet ice
storage temp.
−20°C
suitability
Listeria spp.
InChI
1S/C31H43N3O8/c1-8-12-33-26-21-13-17(2)14-25(41-7)27(36)19(4)15-20(5)29(42-31(32)39)24(40-6)11-9-10-18(3)30(38)34-22(28(21)37)16-23(26)35/h8-11,15-17,19,24-25,27,29,33,36H,1,12-14H2,2-7H3,(H2,32,39)(H,34,38)/b11-9+,18-10-,20-15+/t17-,19-,24-,25-,27+,29-/m1/s1
InChI key
AYUNIORJHRXIBJ-CQFHKJEWSA-N
Related Categories
General description
Biochem/physiol Actions
Hsp90
Packaging
Warning
Reconstitution
Other Notes
Clarke, P.A., et al. 2002. Oncogene19, 4125.
Clarke, P.A., et al. 2000. Oncogene19, 4125.
Schulte, T.W., and Neckers, L.M. 1998. Cancer Chemother. Pharmacol.42, 273.
Schnur, R.C., et al. 1995. J. Med. Chem.38, 3806.
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service