Skip to Content
Merck
All Photos(1)

Key Documents

07-692

Sigma-Aldrich

Anti-Sar1 Antibody

Upstate®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

rabbit

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

monkey, mouse, rat, human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ROR1(4919)

Specificity

Sar1

Immunogen

KLH-conjugated, synthetic peptide corresponding to amino acids 46-62 (CKDDRLGQHVPTLHPTSE) of human Sar1 (a and b isoforms), with an N-terminal cysteine added for conjugation purposes

Application

Detect Sar1 using this Anti-Sar1 Antibody validated for use in WB.
Research Category
Signaling
Research Sub Category
G-proteins

Kinases & Phosphatases

Quality

routinely evaluated by immunoblot on RIPA lysates from 3T3/A31 cells

Target description

Mr 21kDa

Physical form

Format: Purified
Protein A purified

Storage and Stability

stable 2 years at -20°C from date of shipment

Analysis Note

Control
Positive Antigen Control: Catalog #12-305, 3T3/A31 lysate. Add 2.5 μL of 2-mercapto-ethanol/100 μL of lysate and boil for 5 minutes to reduce the preparation. Load 20 μg of reduced lysate per lane for minigels.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bethan Jones et al.
Nature genetics, 34(1), 29-31 (2003-04-15)
Dietary fat is an important source of nutrition. Here we identify eight mutations in SARA2 that are associated with three severe disorders of fat malabsorption. The Sar1 family of proteins initiates the intracellular transport of proteins in COPII (coat protein)-coated
François-Xavier Campbell-Valois et al.
Molecular & cellular proteomics : MCP, 11(7), M111-M111 (2012-03-20)
Phagosomes, by killing and degrading pathogens for antigen presentation, are organelles implicated in key aspects of innate and adaptive immunity. Although it has been well established that phagosomes consist of membranes from the plasma membrane, endosomes, and lysosomes, the notion
A structural view of the COPII vesicle coat.
Bickford, Lincoln C, et al.
Current Opinion in Structural Biology, 14, 147-153 (2004)
Xue-Hai Liang et al.
Nucleic acids research, 46(19), 10225-10245 (2018-09-22)
RNase H1-dependent, phosphorothioate-modified antisense oligonucleotides (PS-ASOs) can enter cells through endocytic pathways and need to be released from the membrane-enclosed organelles, a limiting step for antisense activity. Accumulating evidence has suggested that productive PS-ASO release mainly occurs from late endosomes
The intracellular transport of chylomicrons requires the small GTPase, Sar1b.
Shoulders, Carol C, et al.
Current Opinion in Lipidology, 15, 191-197 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service