Skip to Content
Merck
All Photos(1)

Key Documents

07-1596

Sigma-Aldrich

Anti-SIRT3 (CT) Antibody

from rabbit

Synonym(s):

SIR2-like protein 3 , mitochondrial nicotinamide adenine dinucleotide-dependent deacetylase, silent mating type information regulation 2, S.cerevisiae, homolog 3, sir2-like 3, sirtuin (silent mating type information regulation 2 homolog) 3 (S. cerevisiae

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SIRT3(23410)
mouse ... Sirt3(64384)

General description

SIRT3 is a member of the class I sirtuin family of proteins, homologs to the yeast Sir2 protein. Members of the sirtuin family are characterized by a sirtuin core domain and grouped into four classes. SIRT3 is located in the mitochondria and has been implicated in regulating metabolic processes. Endogenous SIRT3 is a soluble protein located in the mitochondrial matrix. Overexpression of Sirt3 in cultured cells increases respiration and decreases the production of reactive oxygen species. Fasting increases Sirt3 expression in white and brown adipose tissue. A longer form of SIRT3 is present in the nucleus where it exhibits histone deacetylase activity. SIRT3 protects cardiomyocytes from stress mediated cell death due to deacetylation of a nuclear factor, Ku-70.

Specificity

Not tested in other species.
This antibody recognizes SIRT3. Endogenous levels of this protein vary among cell lines; the protein was easily detected in RAW264.7 and C2C12 cells but only weakly detected in HeLa cells, and not detected in Jurkat, HEK293 or PC3 cells.

Immunogen

Epitope: C-terminus
The immunogen was a peptide corresponding the C-terminal region of human SIRT3.

Application

Anti-SIRT3 (C-terminus) Antibody is a Rabbit Polyclonal Antibody for detection of SIRT3 (C-terminus) also known sirtuin 3 & has been validated in WB, ELISA & IHC.
ELISA:
This antibody has been shown by an outside laboratory to be suitable for ELISA (1:1000).

Immunohistochemistry:
This antibody has been shown by an outside laboratory to be suitable for histochemistry using human hepatocarcinoma tissue (1:50 – 1:100).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histone Modifying Proteins

Quality

Western Blot Analysis:
1:250 dilution of this antibody detected SIRT3 on 10 µg of C2C12 lysate.

Target description

44 kDa

Physical form

Format: Purified
Purified
Purified rabbit polyclonal in buffer containing 0.09% NaN3.

Storage and Stability

Stable for 6 months at 2-8°C from date of receipt. For long term storage, store at -20ºC. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
C2C12 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Mar Blanquer-Rosselló et al.
Journal of cellular biochemistry, 114(12), 2785-2794 (2013-07-10)
Xanthohumol (XN) and 8-prenylnaringenin (8PN) are hop (Humulus lupulus L.) polyphenols studied for their chemopreventive effects on certain cancer types. The breast cancer line MCF-7 was treated with doses ranging from 0.001 to 20 µM of XN or 8PN in
Rebecca K Bubenheimer et al.
Frontiers in cellular neuroscience, 10, 73-73 (2016-04-06)
Gut inflammation contributes to the development of gut motility disorders in part by disrupting the function and survival of enteric neurons through mechanisms that involve oxidative stress. How enteric neurons regulate oxidative stress is still poorly understood. Importantly, how neuron
Timothy C Kenny et al.
Cell reports, 27(8), 2292-2303 (2019-05-23)
Moderate mitochondrial stress can lead to persistent activation of cytoprotective mechanisms - a phenomenon termed mitohormesis. Here, we show that mitohormesis primes a subpopulation of cancer cells to basally upregulate mitochondrial stress responses, such as the mitochondrial unfolded protein response (UPRmt)
Jinrong Fu et al.
The Journal of biological chemistry, 287(29), 24460-24472 (2012-06-01)
Huntington disease (HD) is an inherited neurodegenerative disorder caused by an abnormal polyglutamine expansion in the protein Huntingtin (Htt). Currently, no cure is available for HD. The mechanisms by which mutant Htt causes neuronal dysfunction and degeneration remain to be
Dong-Jie Li et al.
British journal of pharmacology, 176(22), 4388-4401 (2018-10-03)
PDGF-BB is an angiogenic factor involved in cardiovascular diseases. Here, we investigated the possible effects of activation of the nicotinic ACh receptor α7 subtype (α7nAChR) on PDGF-BB-induced proliferation and migration in vascular smooth muscle cells (VSMCs). PNU-282987, a selective α7nAChR

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service