Skip to Content
Merck
All Photos(1)

Documents

05-588

Sigma-Aldrich

Anti-HET/SAF-B Antibody, clone 6F7

clone 6F7, Upstate®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6F7, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SAFB(6294)

Specificity

HET/SAF-B

Immunogen

Peptide corresponding to residues 345-357 of human HET/SAF-B

Application

Anti-HET/SAF-B Antibody, clone 6F7 is a high quality Mouse Monoclonal Antibody for the detection of HET/SAF-B & has been validated in IP, WB, ICC & IHC.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

routinely evaluated by immunoblot on whole cell extracts or RIPA lysates from MCF-7 cells

Target description

130kDa

Physical form

0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Format: Purified
Protein G Chromatography

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qimin Zhang et al.
American journal of cancer research, 7(4), 935-945 (2017-05-05)
Bladder cancer is one of the most prevalent cancers worldwide, and increasing research has focused on new technologies for early detection of bladder cancer. For example, proteomic techniques for biomarker discovery have been implemented for the detection and analysis of
S Oesterreich et al.
Molecular endocrinology (Baltimore, Md.), 14(3), 369-381 (2000-03-09)
The estrogen receptor (ER) is a ligand-dependent transcription factor that acts in a cell- and promoter-specific manner. Evidence suggests that the activity of the ER can be regulated by a number of other stimuli (e.g. growth factors) and that the
Caroline Rivers et al.
BMC biology, 13, 111-111 (2015-12-24)
SAFB1 is a RNA binding protein implicated in the regulation of multiple cellular processes such as the regulation of transcription, stress response, DNA repair and RNA processing. To gain further insight into SAFB1 function we used iCLIP and mapped its
E Werwein et al.
Cell death & disease, 4, e511-e511 (2013-03-02)
B-Myb is a highly conserved member of the Myb transcription factor family that has essential roles in cell-cycle progression. Recent work has suggested that B-Myb is also involved in the cellular DNA-damage response. Here, we have investigated the fate of
Khairul I Ansari et al.
Journal of molecular biology, 411(2), 334-349 (2011-06-21)
Homeobox (HOX)-containing gene HOXC6 is a critical player in mammary gland development and milk production, and is overexpressed in breast and prostate cancers. We demonstrated that HOXC6 is transcriptionally regulated by estrogen (E2). HOXC6 promoter contains two putative estrogen response

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service