Skip to Content
Merck
All Photos(3)

Key Documents

04-1481

Sigma-Aldrich

Anti-RPA2 p34 Antibody, clone RPA20 1-46

from mouse, clone RPA20, clone 1-46

Synonym(s):

RF-A protein 2, RP-A p32, RP-A p34, Replication factor A protein 2, replication protein A2 (32kD), replication protein A2, 32kDa, Replication protein A 32kDa subunit, RF-A, replication factor-A protein 2, p32, p34, RPA2, REPA2, RPA32.

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€130.00
100 μG
€377.00

€130.00


Estimated to ship on02 June 2025Details


Request a Bulk Order

Select a Size

Change View
25 μG
€130.00
100 μG
€377.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€130.00


Estimated to ship on02 June 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

1-46, monoclonal
RPA20, monoclonal

species reactivity

human, mouse

packaging

antibody small pack of 25 μg

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... RPA2(6118)

General description

RPA is a heterotrimeric protein complex that binds specifically to single-stranded DNA (ssDNA). It is composed of three subunits, RPA1 (70 kDa), RPA2 (32 kDa), and RPA3 (14 kDa), and plays multiple roles in DNA metabolism. RPA is required for DNA replication initiation, as well as replication elongation. At the onset of DNA replication, RPA is loaded onto chromatin after the binding of Cdc45 to origins. RPA is needed for subsequent loading of DNA polymerase and other replication proteins to initiate DNA replication. After DNA replication begins, RPA moves with replication forks, stabilizing ssDNA and assisting in DNA synthesis. In addition to its replication function, RPA is also known to play essential roles in damage repair and recombination.The 32 kDa subunit, is phosphorylated by the cdc2 family of kinases when cells enter S-phase and in response to DNA damage by ATM, ATR, and DNA-PK. Alternate names for RPA32 include replication protein A 32 kDa subunit, RP-A, RF-A, replication factor-A protein 2, p32, p34, RPA2, REPA2, and RPA32.

Specificity

This antibody recognizes RPA2.

Immunogen

Epitope: Unknown
His-tagged recombinant protein corresponding to human RPA2.

Application

Anti-RPA2 p34 Antibody, clone RPA20 1-46 is a Mouse Monoclonal Antibody for detection of RPA2 p34 also known as Replication factor A protein 2 & has been validated in WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair

Chromatin Biology
Western Blot (SNAP ID) Analysis: 0.5 µg/mL from a previous lot detected RPA2 on 10 µg of NIH/3T3 cell lysate.

Quality

Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected RPA2 on 10 µg of HeLa cell lysate.

Target description

~32 kDa

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lauren M Young et al.
Cell reports, 13(3), 451-459 (2015-10-13)
PARP1 is the main sensor of single- and double-strand breaks in DNA and, in building chains of poly(ADP-ribose), promotes the recruitment of many downstream signaling and effector proteins involved in the DNA damage response (DDR). We show a robust physical
Jang-Won Ahn et al.
Nucleic acids research, 43(13), 6321-6333 (2015-06-13)
DNA double-strand breaks (DSBs) are the most severe type of DNA damage and are primarily repaired by non-homologous end joining (NHEJ) and homologous recombination (HR) in the G1 and S/G2 phase, respectively. Although CtBP-interacting protein (CtIP) is crucial in DNA
Nanda Kumar Sasi et al.
Neoplasia (New York, N.Y.), 20(10), 985-995 (2018-08-30)
CDC7-DBF4 kinase (DDK) initiates DNA replication in eukaryotes by activating the replicative MCM helicase. DDK has diverse and apparently conflicting roles in the replication checkpoint response in various organisms, but the underlying mechanisms are far from settled. We show that
Na Mi Ryu et al.
Cell cycle (Georgetown, Tex.), 18(20), 2660-2671 (2019-08-17)
DNA repair proteins have been found to localize to the centrosomes and defects in these proteins cause centrosome abnormality. Centrobin is a centriole-associated protein that is required for centriole duplication and microtubule stability. A recent study revealed that centrobin is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service