Skip to Content
Merck
All Photos(3)

Key Documents

05-421-AF488

Sigma-Aldrich

Anti-PP2A, C subunit Antibody, clone 1D6, Alexa Fluor 488 Conjugate

clone 1D6, from mouse, ALEXA FLUOR 488

Synonym(s):

Serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform, PP2A-alpha, PP2Ac, PP2Calpha, Replication protein C, RP-C

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€353.00

€353.00


Estimated to ship on17 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€353.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€353.00


Estimated to ship on17 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

purified antibody

antibody product type

primary antibodies

clone

1D6, monoclonal

species reactivity

Xenopus, rabbit, human, bovine, rat, yeast, mouse

technique(s)

immunocytochemistry: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PPP2CA(5515)

General description

Serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform (EC 3.1.3.16; UniProt P67775; PP2A-alpha, PP2Ac, PP2Calpha, Replication protein C, RP-C) is encoded by the PPP2CA (also known as PP2CA) gene (Gene ID 5515) in human. Protein phosphatase 2 (PP2 or PP2A) is a ubiquitously expressed heterotrimeric serine/threonine protein phosphatase with broad substrate specificity, such as Raf, MEK, and AKT. PP2A is composed of a structural A and a catalytic C subunit as the dimeric core enzyme, and a regulatory B subunit. There exist two different A subunits (PPP2R1A & PPP2R1B), two different C subunits (PPP2CA & PPP2CB), and 12 different B subunits (PPP2R2B-D, PPP2R3A-C, PPP2R4, PPP2R5A-E). The enzymatic activity of a catalytic subunit is altered upon binding an A subunit even in the absence of a B subunit. The B subunits are believed to play key roles in controlling the localization and specific activity of the A/C holoenzymes.

Specificity

Expected to recognize both spliced isoforms of human PP2A catalytic subunit reported by UniProt (P67775).

Immunogen

A 16 residue synthetic peptide (C-RGEPHVTRRTPDYFL) corresponding to amino acids 295-309 of the 36 kDa catalytic subunit of human protein phosphatase 2A (PP2A).
Epitope: Amino acids 295-309.

Application

Anti-PP2A, C subunit Antibody, clone 1D6, Alexa Fluor 488 Conjugate is an antibody against PP2A for use in Immunocytochemistry.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected PP2A in NIH/3T3 cells and HUVECs.
The unconjugated antibody (Cat. No. 05-421) is shown to be suitable also for immunocytochemistry, immunoprecipitation, and Western blotting applications.
Research Category
Signaling
Research Sub Category
Kinases & Phosphatases

Quality

Evaluated by Immunocytochemistry in A431 cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected PP2A in A431 cells.

Target description

~36 kDa observed. 35.59 kDa (isoform 1) and 29.74 kDa (isoform 2) calculated.

Physical form

Protein A purified
Purified mouse monoclonal IgG2bκ antibody conjugate in PBS with 15 mg/mL BSA and 0.1 % sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yulia Gryaznova et al.
The EMBO journal, 40(7), e106797-e106797 (2021-03-02)
Partitioning of the genome in meiosis occurs through two highly specialized cell divisions, named meiosis I and meiosis II. Step-wise cohesin removal is required for chromosome segregation in meiosis I, and sister chromatid segregation in meiosis II. In meiosis I

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service